missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DAPI dihydrochloride

Product Code. 16478758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478758 10 mg 10mg
16473816 25 mg 25mg
16454677 50 mg 50mg
16431178 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478758 Supplier MedChemExpress Supplier No. HYD0814
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DAPI dihydrochloride (4',6-diamidino-2-phenylindole) is a fluorescent stain by binding in the minor grove of A-T rich sequences of DNA.

Specifications

Content And Storage -20°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 75 mg/mL (214.13 mM; Need ultrasonic) ∣ H2O : 5.56 mg/mL (15.87 mM; ultrasonic and warming and heat to 60°C)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 350.25 g/mol
CAS 28718-90-3
Synonym 4',6-Diamidino-2-phenylindole dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.