Learn More
GeneBLAzer™ D5-CRE-bla CHO-K1 Cells
GeneBLAzer™ D5-CRE-bla CHO-K1 Cells cells contain the human Dopamine Receptor 5 (D5).
Brand: GeneBLAzer™ K1784
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: DRD5
- Accession Number: NM_000676
- Population Status: Single Cell Clone Origin
- The GeneBLAzer™ D5-CRE-bla CHO-K1 cells contain the human Dopamine Receptor 5 (D5) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. no. K1129) contain a beta-lactamase reporter gene under control of the CRE response element.
- The GeneBLAzer™ D5-CRE-bla CHO-K1 cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, GeneBLAzer™ D5-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Specifications
Liquid Nitrogen | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
D5-CRE-bla CHO-K1 cells, D5-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Dividing Cells |