missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress D149 Dye

Product Code. 16484057
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484057 50 mg 50mg
16471936 500 mg 500mg
16427906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16484057 Supplier MedChemExpress Supplier No. HY50938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D149 Dye is an indoline-based dye, which is a high-extinction-coefficient metal-free organic sensitizer.

Specifications

Quantity 50 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 741.94 g/mol
Solubility DMSO : 1 mg/mL (1.35 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 786643-20-7
Synonym D149 ∣ Indoline dye D149
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.