missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol hexaacetate, 97%, Thermo Scientific™

Product Code. 15437369
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437369 25 g 25g
15447369 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437369 Supplier Thermo Scientific Alfa Aesar Supplier No. J66121.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7208-47-1
Molecular Formula C18H26O12
Molecular Weight (g/mol) 434.394
MDL Number MFCD00008715
InChI Key NJVBTKVPPOFGAT-XMTFNYHQSA-N
Synonym d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
PubChem CID 9802952
IUPAC Name [(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate
SMILES CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 100°C to 104°C
Color White
Quantity 25 g
Formula Weight 434.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material D-Sorbitol hexaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.