accessibility menu, dialog, popup

UserName

CAS RN 7208-47-1

CH3OOOCH3OOH3COOCH3OOCH3OOCH3O

CAS RN 7208-47-1

IUPAC Name: 1,2,4,5,6-pentakis(acetyloxy)hexan-3-yl acetate
Synonyms: 1,2,4,5,6-pentakis(acetyloxy)hexan-3-yl acetate
Molecular Weight (g/mol): 434.39
Molecular Formula: C18H26O12
InChi Key: NJVBTKVPPOFGAT-UHFFFAOYNA-N
SMILES: CC(=O)OCC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C(COC(C)=O)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results