missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Ribose-5-phosphate barium salt hexahydrate, 99%

Product Code. 15467209
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15467209 Supplier Thermo Scientific Chemicals Supplier No. J66017.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15673-79-7 | C5H9BaO8P | 365.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15673-79-7
Molecular Formula C5H9BaO8P
Molecular Weight (g/mol) 365.42
MDL Number MFCD00047534
InChI Key VNFDCDZWBYRPMC-QHEAFLAXSA-L
PubChem CID 123134315
SMILES [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 3781061
Sensitivity Hygroscopic
Optical Rotation +12° (c=1 in 0.2M HCl)
IUPAC Name barium;[(2R,3R,4R)-2,3,4-trihydroxy-5-oxopentyl] dihydrogen phosphate
Formula Weight 473.51
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D-Ribose-5-phosphate barium salt hexahydrate
compliance-icons
Product Identifier
  • D-Ribose-5-phosphate barium salt hexahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: D-Ribose-5-phosphatebarium salt hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.