accessibility menu, dialog, popup

UserName

Ba(S)(R)(R)OOHOHHOOPOOO

CAS RN 15673-79-7

IUPAC Name: barium(2+) (2R,3R,4R)-2,3,4-trihydroxy-5-(phosphonatooxy)pentanal
Synonyms: barium(2+) D-ribose 5-phosphate(2-)
Molecular Weight (g/mol): 365.42
Molecular Formula: C5H9BaO8P
InChi Key: GWEBFOYVTFUYEC-VEGRVEBRSA-L
SMILES: [Ba++].O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@@H](O)C=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)