Learn More
D-(-)-Prolinamide, 98%
Beskrivning
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifikationer
Specifikationer
| Melting Point | 100°C to 103°C |
| Quantity | 5 g |
| Solubility Information | Soluble in water. |
| Optical Rotation | +107° (c=2 in Ethanol) |
| Formula Weight | 114.15 |
| Percent Purity | 98% |
| Chemical Name or Material | D-(-)-Prolinamide |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.