accessibility menu, dialog, popup

UserName

CAS RN 62937-45-5

NHONH2

CAS RN 62937-45-5

IUPAC Name: pyrrolidine-2-carboxamide
Synonyms: pyrrolidine-2-carboxamide
Molecular Weight (g/mol): 114.15
Molecular Formula: C5H10N2O
InChi Key: VLJNHYLEOZPXFW-UHFFFAOYNA-N
SMILES: NC(=O)C1CCCN1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)