missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Penicillamine, 98%, Thermo Scientific™

£53.10 - £585.00

Chemical Identifiers

CAS 52-67-5
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.208
MDL Number MFCD00064302
InChI Key VVNCNSJFMMFHPL-VKHMYHEASA-N
Synonym d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
PubChem CID 5852
ChEBI CHEBI:7959
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460303
SDS View Documents
Thermo Scientific Alfa Aesar
A11446.06
5g
£53.10
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11470303
SDS View Documents
Thermo Scientific Alfa Aesar
A11446.14
25g
£196.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11480303
SDS View Documents
Thermo Scientific Alfa Aesar
A11446.22
100g
£585.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
It is freely soluble in water, slightly soluble in alcohol, insoluble in chloroform, ether, acetone, benzene, and in carbon tetrachloride.

Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C.
Specifications

Chemical Identifiers

52-67-5
149.208
VVNCNSJFMMFHPL-VKHMYHEASA-N
5852
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
C5H11NO2S
MFCD00064302
d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
CHEBI:7959
CC(C)(C(C(=O)O)N)S

Specifications

52-67-5
C5H11NO2S
1722375
d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
VVNCNSJFMMFHPL-VKHMYHEASA-N
CC(C)(C(C(=O)O)N)S
149.208
CHEBI:7959
99%
Unpleasant
MFCD00064302
14,7088
It is freely soluble in water,slightly soluble in alcohol,insoluble in chloroform,ether,acetone,benzene,and in carbon tetrachloride.
−63° (c=1 in 1N NaOH)
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
5852
149.21
D- (-)-Penicillamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P280-P285-P302+P352-P304+P341-P305+P351+P338-P332+P313-P342+P311-P362-P501c

H315-H319-H334

missing translation for 'einecsNumber' : 200-148-8

missing translation for 'rtecsNumber' : YV9425000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.