missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Penicillamine, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A11446.14

Quantity 25g

 View more versions of this product

Product Code. 11470303

  • £196.00 / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
It is freely soluble in water, slightly soluble in alcohol, insoluble in chloroform, ether, acetone, benzene, and in carbon tetrachloride.

Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

52-67-5
149.208
VVNCNSJFMMFHPL-VKHMYHEASA-N
5852
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
C5H11NO2S
MFCD00064302
d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
CHEBI:7959
CC(C)(C(C(=O)O)N)S

Specifications

52-67-5
C5H11NO2S
25g
14,7088
It is freely soluble in water,slightly soluble in alcohol,insoluble in chloroform,ether,acetone,benzene,and in carbon tetrachloride.
−63° (c=1 in 1N NaOH)
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
5852
149.21
D- (-)-Penicillamine
Unpleasant
MFCD00064302
1722375
d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
VVNCNSJFMMFHPL-VKHMYHEASA-N
CC(C)(C(C(=O)O)N)S
149.208
CHEBI:7959
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.