missing translation for 'onlineSavingsMsg'
Learn More

D-Methionine methyl ester hydrochloride, 98%

Product Code. 15430109
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15430109

Brand: Thermo Scientific Alfa Aesar H63422.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69630-60-0 | C6H14ClNO2S | 199.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69630-60-0
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00070385
InChI Key MEVUPUNLVKELNV-NUBCRITNSA-N
Synonym h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
PubChem CID 12888616
IUPAC Name methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CCSC

Specifications

Quantity 100 g
Formula Weight 199.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Methionine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.