Learn More
Thermo Scientific Chemicals D-(+)-Melezitose hydrate, 99%
Description
D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.
Specifications
Specifications
| Melting Point | ∼160°C (decomposition) |
| Quantity | 50 g |
| Beilstein | 99539 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,5819 |
| Solubility Information | Soluble in water. |
| Optical Rotation | +88°(c=4 in Water) |
| Formula Weight | 504.46 (Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | D-(+)-Melezitose hydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.