accessibility menu, dialog, popup

UserName

CAS RN 207511-10-2

(S)(S)(R)(R)(R)OO(S)(S)OOO(R)(S)(S)(R)(R)(R)OHOHHOHOOHHOHOHOHOHOOH

CAS RN 207511-10-2

IUPAC Name: 2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol hydrate
Synonyms: 2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol hydrate
Molecular Weight (g/mol): 522.45
Molecular Formula: C18H34O17
InChi Key: CFXAVQUXSYFPDE-UHFFFAOYNA-N
SMILES: O.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(OC2OC(CO)C(O)C(O)C2O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results