missing translation for 'onlineSavingsMsg'
Learn More

D,L-beta-Chloroalanine Methyl Ester Hydrochloride, TRC

Product Code. 30354485
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354485 2.5 g 2.5g
30359427 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30354485 Supplier Toronto Research Chemicals Supplier No. TRCC3642102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33646-31-0
Molecular Formula C4 H8 Cl N O2 . Cl H
Molecular Weight (g/mol) 174.03
Synonym Alanine, 3-chloro-, methyl ester, hydrochloride (1:1) (ACI), Alanine, 3-chloro-, methyl ester, hydrochloride (6CI, 7CI, 8CI, 9CI), 3-Chloroalanine methyl ester hydrochloride, DL-3-Chloroalanine methyl ester hydrochloride, NSC 170313, NSC 77696, 3-Chloro-DL-alanine methyl ester hydrochloride, β-Chloro-DL-alanine methyl ester hydrochloride, Methyl 2-amino-3-chloropropanoate hydrochloride
IUPAC Name methyl 2-amino-3-chloropropanoate;hydrochloride
SMILES Cl.COC(=O)C(N)CCl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C4H8ClNO2.ClH/c1-8-4(7)3(6)2-5;/h3H,2,6H2,1H3;1H
Formula Weight 173.001
Chemical Name or Material D,L-beta-Chloroalanine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.