CAS RN 33646-31-0
CAS RN 33646-31-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D,L-beta-Chloroalanine Methyl Ester Hydrochloride, TRC
CAS: 33646-31-0 Molecular Formula: C4 H8 Cl N O2 . Cl H Molecular Weight (g/mol): 174.03 Synonym: Alanine, 3-chloro-, methyl ester, hydrochloride (1:1) (ACI),Alanine, 3-chloro-, methyl ester, hydrochloride (6CI, 7CI, 8CI, 9CI),3-Chloroalanine methyl ester hydrochloride,DL-3-Chloroalanine methyl ester hydrochloride,NSC 170313,NSC 77696,3-Chloro-DL-alanine methyl ester hydrochloride,β-Chloro-DL-alanine methyl ester hydrochloride,Methyl 2-amino-3-chloropropanoate hydrochloride IUPAC Name: methyl 2-amino-3-chloropropanoate;hydrochloride SMILES: Cl.COC(=O)C(N)CCl