missing translation for 'onlineSavingsMsg'
Learn More

D-Glucono-1,4-lactone (>85%), TRC

Product Code. 30240627
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
Unit Size:
200mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240627 200 mg 200mg
30240851 50 mg 50mg
30240760 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240627 Supplier Toronto Research Chemicals Supplier No. TRCG417478200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198-69-2
Molecular Formula C6 H10 O6
Molecular Weight (g/mol) 178.14
Synonym D-Gluconic acid, γ-lactone, Gluconic acid, γ-lactone, D- (8CI), 1,4-D-Gluconolactone, 1,4-Gluconolactone, D-Glucono-1,4-lactone, D-Glucono-γ-lactone, D-Gluconolactone, Gluconic acid, γ-lactone, Glucono-γ-lactone
IUPAC Name (3R,4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES OC[C@@H](O)[C@H]1OC(=O)[C@H](O)[C@H]1O

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3-,4-,5-/m1/s1
Formula Weight 178.0477
Chemical Name or Material D-Glucono-1,4-lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.