CAS RN 1198-69-2
CAS RN 1198-69-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-Glucono-1,4-lactone (>85%), TRC
CAS: 1198-69-2 Molecular Formula: C6 H10 O6 Molecular Weight (g/mol): 178.14 Synonym: D-Gluconic acid, γ-lactone,Gluconic acid, γ-lactone, D- (8CI),1,4-D-Gluconolactone,1,4-Gluconolactone,D-Glucono-1,4-lactone,D-Glucono-γ-lactone,D-Gluconolactone,Gluconic acid, γ-lactone,Glucono-γ-lactone IUPAC Name: (3R,4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one SMILES: OC[C@@H](O)[C@H]1OC(=O)[C@H](O)[C@H]1O