missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£155.00
Specifications
| Antigen | D-Dimer |
|---|---|
| Clone | DD1 |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoassay, Sandwich ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
D-Dimer Monoclonal specifically detects D-Dimer in Human, Monkey, Baboon samples. It is validated for Western Blot, ELISA, Immunoassay.Specifications
| D-Dimer | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoassay, Sandwich ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| D Dimer | |
| IgG2a | |
| Protein A purified | |
| is specific for D-Dimer and high molecular weight fibrin degradation products. No cross-reaction with D-Monomer or D-Dimer treated with mercaptoethanol. |
| DD1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer. | |
| Primary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title