missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-8374-0.05mg
This item is not returnable.
View return policy
Description
D-Dimer Monoclonal specifically detects D-Dimer in Human, Monkey, Baboon samples. It is validated for Western Blot, ELISA, Immunoassay.
Specifications
| D-Dimer | |
| Monoclonal | |
| Unconjugated | |
| D Dimer | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer. | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DD1 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoassay, Sandwich ELISA 1:100-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| is specific for D-Dimer and high molecular weight fibrin degradation products. No cross-reaction with D-Monomer or D-Dimer treated with mercaptoethanol. | |
| Store at 4°C. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction