missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Cellobiose octaacetate

Product Code. 11362706
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362706 5 g 5g
11372706 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362706 Supplier Thermo Scientific Chemicals Supplier No. L08780.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5346-90-7 | C28H38O19 | 678.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5346-90-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.59
MDL Number MFCD00009600,MFCD00079432,MFCD00009600,MFCD00078092,MFCD00009600,MFCD00009600
InChI Key WOTQVEKSRLZRSX-UHFFFAOYNA-N
Synonym d-cellobiose octaacetate
PubChem CID 126963588
IUPAC Name [(2R,3R,4R,5R,6S)-4,5,6-triacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 224°C to 231°C
Quantity 5 g
Beilstein 79280
Merck Index 14,1961
Optical Rotation +40° (c=1 in Chloroform)
Formula Weight 678.59
Chemical Name or Material D-Cellobiose octaacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.