CAS RN 5346-90-7
CAS RN 5346-90-7
IUPAC Name:
[4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Synonyms:
[4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Molecular Weight (g/mol):
678.59
Molecular Formula:
C28H38O19
InChi Key:
WOTQVEKSRLZRSX-UHFFFAOYNA-N
SMILES:
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals D-Cellobiose octaacetate
CAS: 5346-90-7 Molecular Formula: C28H38O19 Molecular Weight (g/mol): 678.59 MDL Number: MFCD00009600,MFCD00079432,MFCD00009600,MFCD00078092,MFCD00009600,MFCD00009600 InChI Key: WOTQVEKSRLZRSX-UHFFFAOYNA-N Synonym: d-cellobiose octaacetate PubChem CID: 126963588 IUPAC Name: [(2R,3R,4R,5R,6S)-4,5,6-triacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate SMILES: CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O