missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- D-Biotin for preparation of removal buffer for the use in Streptamer™ technology
- To yield fully functional, highly pure and non-induced cells, fluorescent or magnetic Streptamers™ are efficiently removed from the labeled cells by the addition of biotin
Specifications
Specifications
| Capacity (Metric) | 1 mL |
| Concentration | 100 mM |
| Formulation | D-Biotin |
| Kit Contents | 0.9% (w/w) NaCl |
| pH Range | 7.4 |
| Content And Storage | -25°C to -15°C |
| Quantity | 1 mL |
| For Use With (Application) | Preparation of Removal Buffer for The Use in Streptamer™ Technology |
| Type | Biotin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?