missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Altrose, MP Biomedicals™

Product Code. 11496191 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg

Product Code. 11496191

Brand: MP Biomedicals 0219120810

Please to purchase this item. Need a web account? Register with us today!

Used as a substrate to identify, differentiate and characterize aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1990-29-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-ARQDHWQXSA-N
Synonym d-altro-hexose, 2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexanal, unii-5pln1o36ff, 5pln1o36ff, d-+-altrose, aldehydo-d-altrose
PubChem CID 94780
ChEBI CHEBI:28385
IUPAC Name (2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Color White
Quantity 10 mg
Formula Weight 180.16
Physical Form Powder
Chemical Name or Material D-(+)-Altrose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.