CAS RN 1990-29-0
CAS RN 1990-29-0
IUPAC Name:
2,3,4,5,6-pentahydroxyhexanal
Synonyms:
dextrose corn sugar (14C)-glucose D(+)-glucose (U-14C)glucose (14C)D-glucose sirup D-(U-14C)glucose glucose (D) D(+)-mannose
Molecular Weight (g/mol):
180.16
Molecular Formula:
C6H12O6
InChi Key:
GZCGUPFRVQAUEE-UHFFFAOYNA-N
SMILES:
OCC(O)C(O)C(O)C(O)C=O
1
–
2
of
2
results
1
D-(+)-Altrose, MP Biomedicals™
CAS: 1990-29-0 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 InChI Key: GZCGUPFRVQAUEE-ARQDHWQXSA-N Synonym: d-altro-hexose,2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexanal,unii-5pln1o36ff,5pln1o36ff,d-+-altrose,aldehydo-d-altrose PubChem CID: 94780 ChEBI: CHEBI:28385 IUPAC Name: (2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexanal SMILES: C(C(C(C(C(C=O)O)O)O)O)O