missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ D 4476
GREENER_CHOICE

Product Code. 18033892
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18033892

Brand: Tocris Bioscience™ 2902/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective CK1 inhibitor. Also inhibits TGF-βRI

Selective inhibitor of casein kinase 1 (CK1) and TGF-β type-I receptor (ALK5) that displays > 20-fold selectivity over SAPK2/p38 and a much greater selectivity over all other protein kinases tested. Suppresses site-specific phosphorylation and nuclear exclusion of FOXO1a.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301836-43-1
Molecular Formula C23H18N4O3

Specifications

Quantity 10 mg
Formula Weight 398.41
Purity >99%
Molecular Weight (g/mol) 398.422
Chemical Name or Material D 4476
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.