missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 1C4/SULT1C4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52893
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1C4/SULT1C4 Polyclonal specifically detects Cytosolic Sulfotransferase 1C4/SULT1C4 in Human samples. It is validated for Western Blot.
Specifications
| Cytosolic Sulfotransferase 1C4/SULT1C4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, MGC149521, MGC34422, ST1C4, Sulfotransferase 1C2, sulfotransferase family, cytosolic, 1C, member 4, sulfotransferase family, cytosolic, 1C, member C2, SULT1C#2, SULT1C2cytosolic, 1C, member 2 | |
| Rabbit | |
| 35 kDa | |
| 100 μL | |
| Primary | |
| Porcine: 86%. | |
| Human, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O75897 | |
| SULT1C4 | |
| Synthetic peptides corresponding to SULT1C4(sulfotransferase family, cytosolic, 1C, member 4) The peptide sequence was selected from the middle region of SULT1C4. Peptide sequence HEHVKGWWEAKDKHRILYLFYEDMKKNPKHEIQKLAEFIGKKLDDKVLDK. | |
| Affinity purified | |
| RUO | |
| 27233 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction