missing translation for 'onlineSavingsMsg'
Learn More

Cytosine-5,6-d2, TRC

Product Code. 30355237
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355237 10 mg 10mg
30351462 1 mg 1mg
30348996 2 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30355237 Supplier Toronto Research Chemicals Supplier No. TRCC99895110MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106391-24-6
Molecular Formula C4 2H2 H3 N3 O
Molecular Weight (g/mol) 113.11
Synonym 2(1H)-Pyrimidinone-4,5-d2, 6-amino- (9CI), Cytosine-d2, Cytosine-5,6-D2, 4-Aminopyrimidin-2(1H)-one-5,6-d2
IUPAC Name 4-amino-5,6-dideuterio-1H-pyrimidin-2-one
SMILES [2H]C1=C([2H])C(=NC(=O)N1)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H5N3O/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)/i1D,2D
Formula Weight 113.0558
Chemical Name or Material Cytosine-5,6-D2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.