missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus Antibody (DDG9 and CCH2), Novus Biologicals™
Mouse Monoclonal Antibody
£158.00 - £469.00
Specifications
| Antigen | Cytomegalovirus |
|---|---|
| Clone | DDG9 and CCH2 |
| Dilution | Immunohistochemistry 1:10-1:25, Immunohistochemistry-Paraffin 1:10-1:25 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18054396
|
Novus Biologicals
NBP1-22532SS |
0.1 mL |
£158.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706893
|
Novus Biologicals
NBP1-22532 |
0.5 mL |
£469.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytomegalovirus | |
| Immunohistochemistry 1:10-1:25, Immunohistochemistry-Paraffin 1:10-1:25 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Tissue culture supernatant with 0.05% Sodium Azide | |
| BALB/C mice were immunized with cytomegalovirus infected cell lysate. This antibody is a cocktail of DDG9 and CCH2. | |
| Primary | |
| Store at 4C. Do not freeze. |
| DDG9 and CCH2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cytomegalovirus (CMV), Virus | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| IgG1 κ, IgG2a κ | |
| Tissue culture supernatant | |
| This cocktail antibody reacts with an immediate early and early CMV antigens. The antibody stains infected cells giving a nuclear staining pattern with an immediate early and an early antigen. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title