missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus Antibody (DDG9 and CCH2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22532SS
This item is not returnable.
View return policy
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytomegalovirus | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with 0.05% Sodium Azide | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| This cocktail antibody reacts with an immediate early and early CMV antigens. The antibody stains infected cells giving a nuclear staining pattern with an immediate early and an early antigen. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ, IgG2a κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DDG9 and CCH2 | |
| Immunohistochemistry 1:10-1:25, Immunohistochemistry-Paraffin 1:10-1:25 | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| BALB/C mice were immunized with cytomegalovirus infected cell lysate. This antibody is a cocktail of DDG9 and CCH2. | |
| 0.1 mL | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction