missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Rabbit anti-Human, Clone: MonoPoly/7249R, Novus Biologicals™
Rabbit Monoclonal Antibody
£228.00 - £412.00
Specifications
| Antigen | Cytokeratin, pan |
|---|---|
| Clone | MonoPoly/7249R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369203
|
Bio-Techne
NBP3-13914-20UG |
20 μg |
£228.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003404
|
Novus Biologicals
NBP3-13914-100UG |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Cytokeratin, pan | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant fragments and/or synthetic peptides of human Cytokeratin 76 and Cytokeratin 77 proteins (exact sequences are proprietary) | |
| Primary | |
| Store at 4°C. |
| MonoPoly/7249R | |
| Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 3848 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title