missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Rabbit anti-Human, Clone: MonoPoly/7249R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-13914-100UG
This item is not returnable.
View return policy
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin, pan | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MonoPoly/7249R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant fragments and/or synthetic peptides of human Cytokeratin 76 and Cytokeratin 77 proteins (exact sequences are proprietary) | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 3848 | |
| Store at 4°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction