missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin, pan Antibody (PCK-26), Novus Biologicals™

Mouse Monoclonal Antibody

£170.00

Specifications

Antigen pan Cytokeratin
Clone PCK-26
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300, Immunohistochemistry-Paraffin 1:300 - 1:600, Immunohistochemistry-Frozen 1:300 - 1:600
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18582171
View Documents
Novus Biologicals
NB120-6401-0.025ml
0.025 mL
£170.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Fish, Guinea Pig, Goat, Hamster, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

pan Cytokeratin
Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300, Immunohistochemistry-Paraffin 1:300 - 1:600, Immunohistochemistry-Frozen 1:300 - 1:600
Monoclonal
Ascites
RUO
Human
3848
This Cytokeratin, pan Antibody (PCK-26) was developed against the full length native protein (purified) (Human epidermis).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
PCK-26
Western Blot
Unconjugated
Mouse
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
P04264
KRT1
IgG1
Unpurified
PCK-26 recognizes the 58 kDa cytokeratin 5, the 56 kDa cytokeratin 6 and the 52 kDa cytokeratin 8 band in immunoblotting (Lane EB et al). The antibody recognizes an epitope located on the Type II cytokeratins 1, 5, 6, and 8. PCK-26 is a broad spectrum antibody which reacts specifically with a variety of normal, reactive, and neoplastic epithelial tissues. The antibody reacts with simple, cornifying, and non-cornifying squamous epithelia and pseudostratified epithelia.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.