missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody (PCK-26), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-6401-0.025ml
This item is not returnable.
View return policy
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Fish, Guinea Pig, Goat, Hamster, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| pan Cytokeratin | |
| Monoclonal | |
| Unconjugated | |
| P04264 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| PCK-26 recognizes the 58 kDa cytokeratin 5, the 56 kDa cytokeratin 6 and the 52 kDa cytokeratin 8 band in immunoblotting (Lane EB et al). The antibody recognizes an epitope located on the Type II cytokeratins 1, 5, 6, and 8. PCK-26 is a broad spectrum antibody which reacts specifically with a variety of normal, reactive, and neoplastic epithelial tissues. The antibody reacts with simple, cornifying, and non-cornifying squamous epithelia and pseudostratified epithelia. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| PCK-26 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300, Immunohistochemistry-Paraffin 1:300 - 1:600, Immunohistochemistry-Frozen 1:300 - 1:600 | |
| KRT1 | |
| This Cytokeratin, pan Antibody (PCK-26) was developed against the full length native protein (purified) (Human epidermis). | |
| 0.025 mL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 3848 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction