missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£181.00 - £361.00
Specifications
| Antigen | pan Cytokeratin |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:75 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018903
|
Novus Biologicals
NB600-579SS |
0.1 mL |
£181.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18785913
|
Novus Biologicals
NB600-579 |
0.5 mL |
£361.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin, pan Polyclonal specifically detects Cytokeratin, pan in Human, Mouse, Bovine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| pan Cytokeratin | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| KRT1 | |
| IgG | |
| Protein A purified | |
| NB600-579 has cross-reactivity with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa molecular weight. The antibody is well suited for the staining of a broad spectrum of human keratins. |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:75 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Bovine, Monkey | |
| 3848 | |
| This Cytokeratin, pan Antibody was developed against pan Cytokeratin isolated from bovine muzzle epidermis. | |
| Primary | |
| Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title