missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, pan Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB600-579SS
This item is not returnable.
View return policy
Description
Cytokeratin, pan Polyclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Bovine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| pan Cytokeratin | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:75 - 1:200 | |
| KRT1 | |
| This Cytokeratin, pan Antibody was developed against pan Cytokeratin isolated from bovine muzzle epidermis. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 3848 | |
| Human, Mouse, Bovine, Monkey | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB600-579 has cross-reactivity with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa molecular weight. The antibody is well suited for the staining of a broad spectrum of human keratins. | |
| Store at 4C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction