missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 8 Ab-4 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunoprecipitation experiments with Epredia™ Cytokeratin 8 Ab-4, Mouse Monoclonal Antibody.

£96.16 - £460.00

Specifications

Antigen Cytokeratin 8 Ab-4
Clone TS1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Host Species Mouse
View More Specs

Products 9
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
12662637
View Documents
Epredia
MS-997-B0
100 μL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12652637
View Documents
Epredia
MS-997-B1
500 μL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12632637
View Documents
Epredia
MS-997-B
1 mL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12682637
View Documents
Epredia
MS-997-P0
100 μL Unconjugated Concentrated 200µg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12602647
View Documents
Epredia
MS-997-P1ABX
100 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12612647
View Documents
Epredia
MS-997-PABX
200 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12692637
View Documents
Epredia
MS-997-P1
500 μL Unconjugated Concentrated 200µg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12672637
View Documents
Epredia
MS-997-P
1 mL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12642647
View Documents
Epredia
MS-997-R7
7 mL Unconjugated Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with keratin 18. Keratin 8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin polypeptides 8 and 18.

Host Species: Mouse

Clone: TS1

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: aa 343-357

Immunogen: Keratin preparation from a human carcinoma

Molecular Weight: 52.5 kDa

Positive Control: MCF-7 or A431 cells; Skin; Colon, Lung or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation

Specifications

Specifications

Cytokeratin 8 Ab-4
Immunohistochemistry (Paraffin), Immunoprecipitation
Mouse
Cytoskeletal Protein
Keratin preparation from a human carcinoma
Primary
TS1
Monoclonal
IVD
Human
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.