missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 8 Ab-4 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunoprecipitation experiments with Epredia™ Cytokeratin 8 Ab-4, Mouse Monoclonal Antibody.

Brand:  Epredia MS-997-P1

 View more versions of this product

Product Code. 12692637

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with keratin 18. Keratin 8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin polypeptides 8 and 18.

Host Species: Mouse

Clone: TS1

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: aa 343-357

Immunogen: Keratin preparation from a human carcinoma

Molecular Weight: 52.5 kDa

Positive Control: MCF-7 or A431 cells; Skin; Colon, Lung or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 8 Ab-4
Monoclonal
200µg/mL
Mouse
500 μL
Cytoskeletal Protein
Human
IgG2a κ
Immunohistochemistry (Paraffin), Immunoprecipitation
TS1
Unconjugated
Keratin preparation from a human carcinoma
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.