missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 75 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55236
This item is not returnable.
View return policy
Description
Cytokeratin 75 Polyclonal specifically detects Cytokeratin 75 in Human samples. It is validated for Western Blot.
Specifications
| Cytokeratin 75 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CK-75, Cytokeratin-75, hK6hf, K6HFcytokeratin type II, K75, KB18, keratin 75, keratin, type II cytoskeletal 75, Keratin-6 hair follicle, Keratin-75, PFB, type II keratin-18, Type II keratin-K6hf, Type-II keratin Kb18 | |
| Rabbit | |
| 59 kDa | |
| 100 μL | |
| Primary | |
| Mouse: 86%; Porcine: 85%; Bovine: 79%; . | |
| Human, Mouse, Rat, Pig, Bovine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O95678 | |
| KRT75 | |
| Synthetic peptides corresponding to Cytokeratin 75 The peptide sequence was selected from the N terminal of Cytokeratin 75. Peptide sequence MSRQSSITFQSGSRRGFSTTSAITPAAGRSRFSSVSVARSAAGSGGLGRI. | |
| Affinity purified | |
| RUO | |
| 9119 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction