Learn More
Epredia™ Lab Vision™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody.
£194.00 - £872.41
Specifications
| Antigen | Cytokeratin 5/6 Ab-2 |
|---|---|
| Clone | D5/16 B4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12670197
|
Epredia™
MS-1814-S0 |
100 μL | Concentrated | ||||||
|
12660197
|
Epredia™
MS-1814-S1 |
500 μL | Concentrated |
£820.00 £778.09 / 500µL Save £41.91 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12650197
|
Epredia™
MS-1814-S |
1 mL | Concentrated |
£922.00 £872.41 / 1mL Save £49.59 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12630197
|
Epredia™
MS-1814-R7 |
7 mL | Prediluted |
£520.00 £489.92 / 7mL Save £30.08 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12640197
|
Epredia™
MS-1814-RQ |
12 mL | Ready-to-Use |
£693.00 £655.49 / 12mL Save £37.51 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification.Host Species: Mouse
Clone: D5/16 B4
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Isolated cytokeratin 5
Molecular Weight: 58 kDa (Cytokeratin 5), 56 kDa (Cytokeratin 6)
Positive Control: Mesothelioma, Squamous Cell Ca
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Cytokeratin 5/6 Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Isolated cytokeratin 5 |
| D5/16 B4 | |
| Monoclonal | |
| Mouse | |
| Cytoskeletal Protein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.