missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1814-R7

 View more versions of this product

Product Code. 12630197

  • ONLINE EXCLUSIVE PRICE
    £524.00
    £489.92 / 7mL
    Save £34.08
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification.

Host Species: Mouse

Clone: D5/16 B4

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Isolated cytokeratin 5

Molecular Weight: 58 kDa (Cytokeratin 5), 56 kDa (Cytokeratin 6)

Positive Control: Mesothelioma, Squamous Cell Ca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 5/6 Ab-2
Monoclonal
Unconjugated
Isolated cytokeratin 5
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
D5/16 B4
Mouse
7 mL
Cytoskeletal Protein
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.