missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 3, Mouse anti-Human, Clone: KRT3/2130, Novus Biologicals™
Mouse Monoclonal Antibody
£204.00 - £377.00
Specifications
| Antigen | Cytokeratin 3 |
|---|---|
| Clone | KRT3/2130 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18005406
|
Novus Biologicals
NBP2-79718-20UG |
20 μg |
£204.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246487
|
Novus Biologicals
NBP2-79718-100UG |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cytokeratin 3 | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| P12035 | |
| 3850 | |
| Recombinant full-length human Cytokeratin 3 protein | |
| Primary | |
| Store at 4°C. |
| KRT3/2130 | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
| KRT3 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title