missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 3, Mouse anti-Human, Clone: KRT3/2130, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79718-20UG
This item is not returnable.
View return policy
Description
Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Cytokeratin 3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P12035 | |
| KRT3 | |
| Recombinant full-length human Cytokeratin 3 protein | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| KRT3/2130 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3850 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction