missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 20 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 20 Ab-1, Mouse Monoclonal Antibody.

£133.00 - £506.65

Specifications

Antigen Cytokeratin 20 Ab-1
Clone Ks20.8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12631237
View Documents
Epredia™
MS-377-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12641237
View Documents
Epredia™
MS-377-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
£355.00
£333.88 / 500µL
Save £21.12
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12621237
View Documents
Epredia™
MS-377-S
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
£539.00
£506.65 / 1mL
Save £32.35
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12611237
View Documents
Epredia™
MS-377-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
£263.00
£246.29 / 7mL
Save £16.71
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12601237
View Documents
Epredia™
MS-377-RQ
12 mL Ready-to-Use
ONLINE EXCLUSIVE PRICE
£349.00
£327.92 / 12mL
Save £21.08
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Keratin 20/cytokeratin20 (CK20) is a Type-I keratin which is primarily expressed in gastric and intestinal epithelium, urothelium, and Merkel-cells. CK20 is expressed in adenocarcinomas of the colon, stomach, pancreas and the bile system. CK20 is also present in mucinous ovarian tumors, transitional-cell and Merkel-cell carcinomas. Notably, the squamous cell carcinomas and adenocarcinomas of the breast, lung, and endometrium, non-mucinous tumors of the ovary, and small cell carcinomas lack in CK20.

Host Species: Mouse

Clone: Ks20.8

Isotype: IgG2a

Species Reactivity: Human and Mouse. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human cytokeratin preparation

Molecular Weight: 46 kDa

Positive Control: HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Specifications

Cytokeratin 20 Ab-1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IVD
Semi-purified human cytokeratin preparation
Ks20.8
Monoclonal
Mouse
Cytoskeletal Protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.