missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 20 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 20 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-377-R7

 View more versions of this product

Product Code. 12611237

  • ONLINE EXCLUSIVE PRICE
    £263.00
    £246.29 / 7mL
    Save £16.71
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keratin 20/cytokeratin20 (CK20) is a Type-I keratin which is primarily expressed in gastric and intestinal epithelium, urothelium, and Merkel-cells. CK20 is expressed in adenocarcinomas of the colon, stomach, pancreas and the bile system. CK20 is also present in mucinous ovarian tumors, transitional-cell and Merkel-cell carcinomas. Notably, the squamous cell carcinomas and adenocarcinomas of the breast, lung, and endometrium, non-mucinous tumors of the ovary, and small cell carcinomas lack in CK20.

Host Species: Mouse

Clone: Ks20.8

Isotype: IgG2a

Species Reactivity: Human and Mouse. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human cytokeratin preparation

Molecular Weight: 46 kDa

Positive Control: HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 20 Ab-1
Monoclonal
Unconjugated
Semi-purified human cytokeratin preparation
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin), Western Blot
Ks20.8
Mouse
7 mL
Cytoskeletal Protein
Human, Mouse
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.