missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Rabbit anti-Human, Clone: 9O6X9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Cytokeratin 17 |
|---|---|
| Clone | 9O6X9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319332
|
Novus Biologicals
NBP3-16271-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303193
|
Novus Biologicals
NBP3-16271-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Western BlotSpecifications
| Cytokeratin 17 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3872 | |
| IgG | |
| Affinity purified |
| 9O6X9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cytokeratin 17 (Q04695). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel