missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Rabbit anti-Human, Clone: 9O6X9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16271-100UL
This item is not returnable.
View return policy
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Western Blot
Specifications
| Cytokeratin 17 | |
| Monoclonal | |
| Unconjugated | |
| 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cytokeratin 17 (Q04695). | |
| 100 μg | |
| Cancer | |
| 3872 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 9O6X9 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction