missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Antibody (KS-1A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£147.00 - £547.00
Specifications
| Antigen | Cytokeratin 13 |
|---|---|
| Clone | KS-1A3 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18041486
|
Novus Biologicals
NBP1-22777-0.25ML |
0.25 mL |
£147.00
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18266350
|
Novus Biologicals
NBP1-22777 |
1 mL |
£547.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 13 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
| KRT13 | |
| IgG1 | |
| Ascites | |
| This is specific to a 54 kD protein. Cytokeratin 13 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 4. Cytokeratin 13 is expressed in major components of squamous, non-keratinized epithelium, transitional epithelium, and myoepithelium. This antibody is useful for identifying carcinomas of trachea, sweat glands, bladder, extocervix, tongue, esophagus, anal canal, and the basal layer of keratinized epidermis. |
| KS-1A3 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 3860 | |
| Cultured A431 cells from a human epidermoid carcinoma of the vulva. | |
| Primary | |
| Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title