missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Antibody (KS-1A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-22777
This item is not returnable.
View return policy
Description
Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 13 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.05% Sodium Azide | |
| KRT13 | |
| Cultured A431 cells from a human epidermoid carcinoma of the vulva. | |
| 1 mL | |
| Cytoskeleton Markers | |
| 3860 | |
| Human | |
| Ascites |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| KS-1A3 | |
| Flow Cytometry, Immunohistochemistry 1:25-1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:25-1:100 | |
| CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| This is specific to a 54 kD protein. Cytokeratin 13 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 4. Cytokeratin 13 is expressed in major components of squamous, non-keratinized epithelium, transitional epithelium, and myoepithelium. This antibody is useful for identifying carcinomas of trachea, sweat glands, bladder, extocervix, tongue, esophagus, anal canal, and the basal layer of keratinized epidermis. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction