missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (18B6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Cytokeratin 10 |
|---|---|
| Clone | 18B6 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025599
|
Novus Biologicals
NBP3-26178-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30035256
|
Novus Biologicals
NBP3-26178-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Cytokeratin 10 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis, keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| A synthesized peptide derived from Human Cytokeratin 10 [UniProt P13645] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 18B6 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3858 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title